Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT7 Mouse anti-Human, Clone: K72.7, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14309

Catalog No. 89-034-391


Add to cart

Description

Description

Mouse monoclonal antibody raised against native human KRT7.

  • High specificity
  • High reactivity
Specifications

Specifications

KRT7
K72.7
Mouse monoclonal antibody raised against native human KRT7.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Liquid
KRT7
Semi-purified cytokeratin preparation.
Protein A/G purification
RUO
Primary
3855
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
P08729
CK7|K2C7|K7|MGC129731|MGC3625|SCL
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)