Learn More
KRT7 Mouse anti-Human, Clone: OTI3E5, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF803333
Catalog No. 50-166-6434
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Blocks interferon-dependent interphase and stimulates DNA synthesis in cells. Involved in the translational regulation of the human papillomavirus type 16 E7 mRNA (HPV16 E7).
Specifications
Specifications
KRT7
Monoclonal
Unconjugated
KRT7
Lyophilized
KRT7
Human recombit protein fragment corresponding to amino acids 1-114 and 340-469 of human KRT7 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI3E5
PBS with 8% trehalose and no preservative; pH 7.3
P08729
CK7, K2C7, K7, SCL
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3855