Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KRT74 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545864
Description
Peptide sequence: HPSSAGVDLG ASAVAGSSGS TQSGQTKTTE ARGGDLKDTQ GKSTPASIPA Sequence homology: Human: 100%.
Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This protein belongs to a family of keratins that are specifically expressed in the inner root sheath of hair follicles.
Specifications
KRT74 | |
Polyclonal | |
Unconjugated | |
KRT74 | |
ADWH; CK-74; cytokeratin-74; HTSS2; HYPT3; K5C; K6IRS4; K74; KB37; keratin 5c; keratin 6 irs4; keratin 74; keratin 74, type II; keratin, type II cytoskeletal 74; Keratin-5c; keratin-74; KRT5C; KRT6IRS4; KRT74; Type II inner root sheath-specific keratin-K6irs4; type-II keratin Kb37 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
121391 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q7RTS7 | |
KRT74 | |
Synthetic peptide directed towards the C-terminal of human KRT74 (aa 475-524). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction