Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRT76 Mouse anti-Human, Rat, Clone: KRTH/1076, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14645
Description
Mouse monoclonal antibody raised against full length recombinant human KRT76.
- High specificity
- High reactivity
Specifications
KRT76 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined | |
Q01546 | |
KRT76 | |
Recombinant protein corresponding to full length human KRT76. | |
100 μg | |
Primary | |
This antibody recognizes basic (Type II or HMW) cytokeratins, which include 67 kDa (CK1); 64 kDa (CK3); 59 kDa (CK4); 58 kDa (CK5); 56 kDa (CK6); 52 kDa (CK8). | |
Store at 4°C. | |
IgG1 κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
KRTH/1076 | |
Mouse monoclonal antibody raised against full length recombinant human KRT76. | |
In 10 mM PBS (0.05% BSA and 0.05% azide). | |
HUMCYT2A/KRT2B/KRT2P | |
Mouse | |
Protein A/G purification | |
RUO | |
51350 | |
Human, Rat | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction