Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KRT77 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570731
Description
This target displays homology in the following species: Cow: 92%; Dog: 92%; Horse: 92%; Human: 100%; Mouse: 93%; Pig: 92%; Rabbit: 92%; Rat: 93%; Sheep: 92%.
Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. KRT77 encodes an epithelial keratin that is expressed in the skin and eccrine sweat glands. The type II keratins are clustered in a region of chromosome 12q13.
Specifications
KRT77 | |
Polyclonal | |
Unconjugated | |
KRT77 | |
4732484G22Rik; CK-1B; CK-2P; cytokeratin 2; cytokeratin-1B; Cytokeratin-2P; embryonic type II keratin 1; embryonic type II keratin-1; HUMCYT2A; K1B; K1emb; K2P; K76; K77; Kb39; keratin 1b; keratin 2p; keratin 76; keratin 76, type II; keratin 77; keratin 77, type II; keratin complex 2, basic, gene 39; keratin, type II cytoskeletal 1b; Keratin, type II cytoskeletal 2 oral; keratin-76; Keratin-77; Krt1b; Krt239; KRT2B; KRT2P; KRT76; Krt77; mK1b; type II keratin Kb39; type-II intermediate filament protein; type-II keratin Kb39; Type-II keratin Kb9 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
374454 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q7Z794 | |
KRT77 | |
synthetic peptide directed towards the C-terminal region of human KRT77. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction