Learn More
KRT8 293T Cell Overexpression Lysate 1 (Denatured), Abnova
Manufacturer:  Abnova Corporation H00003856T01
Catalog No. 89-009-197
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
293T
BC000654
6.8
Transient overexpression cell Lysate was tested with Anti-KRT8 antibody (H00003856-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
KRT8 293T Cell Transient Overexpression Lysate(Denatured)
53.24
pCMV-KRT8 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot