Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT8/KRT18 Mouse anti-Human, Clone: B22.1/B23.1, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14668

Catalog No. 89-034-751


Add to cart

Description

Description

Mouse monoclonal antibody raised against native human KRT8 and KRT18.

  • High specificity
  • High reactivity
Specifications

Specifications

KRT8/KRT18
B22.1/B23.1
Mouse monoclonal antibody raised against native human KRT8 and KRT18.
In PBS.
Liquid
KRT8
A mixture of cytoskeletal preparation from HeLa and PMC-42 cells.
Protein G purification
RUO
Primary
3856|3875
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by
P05787; P05783
CARD2|CK8|CYK8|K2C8|K8|KO
Mouse
IgG
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)