Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT8/KRT18 Mouse anti-Human, Clone: K8.8 + DC10, Abnova™

Mouse monoclonal antibody raised against human KRT8/KRT18.

Manufacturer:  Abnova Corporation MAB12741

Catalog No. 89-934-447


Add to cart

Description

Description

KRT18 encodes the type I intermediate filament chain keratin 18. Keratin 18, together with its filament partner keratin 8, are perhaps the most commonly found members of the intermediate filament gene family. They are expressed in single layer epithelial tissues of the body. Mutations in this gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Specifications

Specifications

KRT8/KRT18
K8.8 + DC10
Mouse monoclonal antibody raised against human KRT8/KRT18.
In 10mM PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide).
CARD2/CK8/CYK8/K2C8/K8/KO
Mouse
Protein G purification
RUO
Primary
3856/3875
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
KRT8
Semi-purified human cytokeratin preparation.
250 uL
Store at 4°C. Do not freeze.
Monoclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit