Learn More
KRT8/KRT18 Mouse anti-Human, Clone: SPM141, Abnova™
Mouse monoclonal antibody raised against native human KRT8/KRT18.
Manufacturer:  Abnova Corporation MAB13213
Catalog No. 89-934-448
Add to cart
Description
Description
KRT18 encodes the type I intermediate filament chain keratin 18. Keratin 18, together with its filament partner keratin 8, are perhaps the most commonly found members of the intermediate filament gene family. They are expressed in single layer epithelial tissues of the body. Mutations in this gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]
Specifications
Specifications
KRT8/KRT18
SPM141
Mouse monoclonal antibody raised against native human KRT8/KRT18.
In 10mM PBS (0.05% BSA, 0.05% sodium azide).
CARD2/CK8/CYK8/K2C8/K8/KO
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determ
Liquid
KRT8
Keratin preparation from human breast carcinoma MCF-7 cells.
Protein A/G purification
RUO
Primary
3856/3875