Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT8 Mouse anti-Human, Clone: 35-beta-H11, Abnova™

Mouse monoclonal antibody raised against human KRT8.

Manufacturer:  Abnova Corporation MAB12675

Catalog No. 89-934-442

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells. The product of this gene typically dimerizes with keratin 18 to form an intermediate filament in simple single-layered epithelial cells. This protein plays a role in maintaining cellular structural integrity and also functions in signal transduction and cellular differentiation. Mutations in this gene cause cryptogenic cirrhosis. [provided by RefSeq]

Specifications

Specifications

KRT8
35-beta-H11
Mouse monoclonal antibody raised against human KRT8.
In PBS (0.09% sodium azide).
CARD2/CK8/CYK8/K2C8/K8/KO
Mouse
IgM
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry Immunocytochemistry Immunohistochemistry (Frozen sections) (1:100-1:500) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:100-1:1000) The optimal working dilution should be deter
Liquid
KRT8
Cytoskeletal preparation of the human hepotocellular carcinoma cell line Hep3B.
Protein G purification
RUO
Primary
3856
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit