Learn More
KRT8 Mouse anti-Human, Clone: M20, Abnova™
Mouse monoclonal antibody raised against human KRT8.
Manufacturer:  Abnova Corporation MAB12669
Catalog No. 89-934-445
Add to cart
Description
Description
This gene is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells. The product of this gene typically dimerizes with keratin 18 to form an intermediate filament in simple single-layered epithelial cells. This protein plays a role in maintaining cellular structural integrity and also functions in signal transduction and cellular differentiation. Mutations in this gene cause cryptogenic cirrhosis. [provided by RefSeq]
Specifications
Specifications
KRT8
M20
Mouse monoclonal antibody raised against human KRT8.
In PBS (0.09% sodium azide).
CARD2/CK8/CYK8/K2C8/K8/KO
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry Immunocytochemistry Immunohistochemistry (Frozen sections) (1:100-1:200 ) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:100-1:1000) The optimal working dilution should be dete
Liquid
KRT8
Isolated from the human breast carcinoma cell line MCF-7.
Protein G purification
RUO
Primary
3856