Learn More
Abnova™ KRT81 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00003887T02
Catalog No. 89-950-258
Add to cart
Description
Description
Specifications
Specifications
293T
BC021241.2
pCMV-KRT81 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
KRT81 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 21.1kDa
Transient overexpression cell lysate was tested with anti-KRT81 antibody (Mfr. No. H00003887-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human