Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KRT85 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570732
Description
This target displays homology in the following species: Cow: 92%; Dog: 86%; Guinea Pig: 86%; Horse: 92%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 86%; Rat: 86%; Sheep: 92%.
KRT85 gene ontology annotations related to this gene include structural molecule activity.
Specifications
KRT85 | |
Polyclonal | |
Unconjugated | |
KRT85 | |
AA589597; BB007925; ECTD4; hair keratin; Hair keratin K2.12; hard keratin, type II, 5; HB5; Hb-5; hHb5; K85; Kb25; keratin 85; keratin 85, type II; keratin 89; keratin complex 2, basic, gene 18; keratin, hair, basic, 5; keratin, type II cuticular Hb5; Keratin-85; Krt2-18; Krt85; KRT89; Krthb5; Type II hair keratin Hb5; type II keratin Kb25; type-II keratin Kb25 | |
Rabbit | |
Affinity chromatography | |
RUO | |
3891 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P78386 | |
KRT85 | |
synthetic peptide directed towards the N-terminal region of human KRT85. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction