Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRTAP2-4 Mouse anti-Human, Clone: OTI3B4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525738

Catalog No. PIMA525738


Add to cart

Description

Description

In the hair cortex, hair keratin intermediate filaments are embedded in an interfilamentous matrix, consisting of hair keratin-associated proteins (KRTAP), which are essential for the formation of a rigid and resistant hair shaft through their extensive disulfide bond cross-linking with abundant cysteine residues of hair keratins. The matrix proteins include the high-sulfur and high-glycine-tyrosine keratins.
Specifications

Specifications

KRTAP2-4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BYR9
KAP2.4; KRTAP2.4
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
85294
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI3B4
Unconjugated
KRTAP2-4
Liquid
KRTAP2-4
Human recombit protein fragment corresponding to amino acids 1-128 of KRTAP2 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit