Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRTAP2 Mouse anti-Human, Clone: OTI3B4, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA503766
Description
In the hair cortex, hair keratin intermediate filaments are embedded in an interfilamentous matrix, consisting of hair keratin-associated proteins (KRTAP), which are essential for the formation of a rigid and resistant hair shaft through their extensive disulfide bond cross-linking with abundant cysteine residues of hair keratins. The matrix proteins include the high-sulfur and high-glycine-tyrosine keratins.Specifications
KRTAP2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9BYR9 | |
KAP2.1B, KAP2.4, KRTAP2.4 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
85294 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI3B4 | |
Unconjugated | |
KRTAP2-4 | |
Liquid | |
KRTAP2-4 | |
Human recombit protein fragment corresponding to amino acids 1-128 of human KRTAP2 produced in E.coli. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title