Learn More
Abnova™ KRTHA2 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00003882T02
Catalog No. 89-950-287
Add to cart
Specifications
Specifications
293T
BC148740
pCMV-KRTHA2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
KRTHA2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 49.28kDa
Transient overexpression cell lysate was tested with anti-KRTHA2 antibody (Mfr. No. H00003882-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human