Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Ku (p70) Ab-4, Mouse Monoclonal Antibody

Recommended for multiple molecular biology techniques, Thermo Scientific™ Ku (p70) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer:  Thermo Scientific™ MS329R7

 View more versions of this product

Catalog No. MS329R7


Add to cart

Description

Description

Recommended for multiple molecular biology techniques, Thermo Scientific™ Ku (p70) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results. Mouse Monoclonal Antibody

The Ku autoantigen is a heterodimer of 70kDa (p70) and ~80kDa (p80) proteins. The p70/p80 dimer is important for function of a 460kDa DNA-dependent protein kinase that phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis.

Host Species: Mouse

Clone: N3H10

Isotype: IgG2b

Species Reactivity: Human, Monkey and Xenopus. Mouse and Hamster (Western Blotting only). Does not react with cow, rabbit and sea urchin.

Epitope: aa 506-541

Immunogen: Human placental extract designated as PSE1-PL

Molecular Weight: 70kDa

Positive Control: BT474, T47D, HEP-G-2 or HeLa cells; Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Ku (p70) Ab-4
Monoclonal
Unconjugated
Prediluted
Human placental extract designated as PSE1-PL
7mL
DNA Repair
Primary
Human, Non-human Primate, Xenopus
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
N3H10
Ready-to-use for IHC (UVLP)
Murine
IgG2b
RUO
Primary
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit