Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Ku (p70) Ab-4, Mouse Monoclonal Antibody

Recommended for multiple molecular biology techniques, Thermo Scientific™ Ku (p70) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

$482.50 - $910.00

Specifications

Antigen Ku (p70) Ab-4
Clone N3H10
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS329P
View Documents
Thermo Scientific™
MS329P
Purified Ab with BSA and Azide 1mL
Each for $699.00
Add to cart
 
MS329P1ABX
View Documents
Thermo Scientific™
MS329P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $482.50
Add to cart
 
MS329PABX
View Documents
Thermo Scientific™
MS329PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $910.00
Add to cart
 
MS329R7
View Documents
Thermo Scientific™
MS329R7
Ready-to-use for IHC (UVLP) 7mL
Each for $512.00
Add to cart
 
MS329R1
View Documents
Thermo Scientific™
MS329R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
MS329P0
View Documents
Thermo Scientific™
MS329P0
Purified Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS329P1
View Documents
Thermo Scientific™
MS329P1
Purified Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Recommended for multiple molecular biology techniques, Thermo Scientific™ Ku (p70) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results. Mouse Monoclonal Antibody

The Ku autoantigen is a heterodimer of 70kDa (p70) and ~80kDa (p80) proteins. The p70/p80 dimer is important for function of a 460kDa DNA-dependent protein kinase that phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis.

Host Species: Mouse

Clone: N3H10

Isotype: IgG2b

Species Reactivity: Human, Monkey and Xenopus. Mouse and Hamster (Western Blotting only). Does not react with cow, rabbit and sea urchin.

Epitope: aa 506-541

Immunogen: Human placental extract designated as PSE1-PL

Molecular Weight: 70kDa

Positive Control: BT474, T47D, HEP-G-2 or HeLa cells; Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Ku (p70) Ab-4
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RUO
Primary
IgG2b
N3H10
Monoclonal
Murine
DNA Repair
Human placental extract designated as PSE1-PL
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit