Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Ku (p80) Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in a variety of molecular biology techniques with the Thermo Scientific™ Ku (p80) Ab-2, Mouse Monoclonal Antibody..

Specifications

Antigen Ku (p80) Ab-2
Clone 111
Applications Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Format Concentrated
View More Specs

Products 2
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS285P0
View Documents
Thermo Scientific™
MS285P0
Purified Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS285P1
View Documents
Thermo Scientific™
MS285P1
Purified Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in a variety of molecular biology techniques with the Thermo Scientific™ Ku (p80) Ab-2, Mouse Monoclonal Antibody.

The Ku autoantigen is a heterodimer of 70kDa (p70) and ~80kDa (p80) proteins. The p70/p80 dimer is important for function of a 460kDa DNA-dependent protein kinase that phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis.

Host Species: Mouse

Clone: 111

Isotype: IgG1/κ

Species Reactivity: Human and Monkey. Does not react with cow, rabbit, mouse and rat.

Epitope: aa 610-705

Immunogen: Human B-cell nuclei from plasmacytoid 2p68 cells

Molecular Weight: 80kDa

Positive Control: HeLa, T47D or BT474 cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Ku (p80) Ab-2
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
Concentrated
RUO
Primary
IgG2a kappa
111
Monoclonal
Murine
DNA Repair
Human B cell nuclei from plasmacytoid 2p68 cells
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit