Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KU14R, Tocris Bioscience™

Catalog Number 113110
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
189224-48-4
C13H14N2O
214.268
JCWVNNMJXQJVNC-UHFFFAOYSA-N
ku14r, 2-2-ethyl-2,3-dihydro-2-benzofuranyl-1h-imidazole, 2-2-ethyl-3h-1-benzofuran-2-yl-1h-imidazole, 1h-imidazole,2-2-ethyl-2,3-dihydro-2-benzofuranyl, 1h-imidazole, 2-2-ethyl-2,3-dihydro-2-benzofuranyl, ku-14r, 2-2-ethyl-2,3-dihydrobenzofuran-2-yl-1h-imidazole, 2-2-ethyl-2,3-dihydro-2-benzo b furanyl-1h-imidazole, 2-2-ethyl-2,3-dihydrobenzo b furan-2-yl-1h-imidazole
4302964
2-(2-ethyl-3H-1-benzofuran-2-yl)-1H-imidazole
CCC1(CC2=CC=CC=C2O1)C3=NC=CN3
This item is not returnable. View return policy
189224-48-4
C13H14N2O
214.268
JCWVNNMJXQJVNC-UHFFFAOYSA-N
ku14r, 2-2-ethyl-2,3-dihydro-2-benzofuranyl-1h-imidazole, 2-2-ethyl-3h-1-benzofuran-2-yl-1h-imidazole, 1h-imidazole,2-2-ethyl-2,3-dihydro-2-benzofuranyl, 1h-imidazole, 2-2-ethyl-2,3-dihydro-2-benzofuranyl, ku-14r, 2-2-ethyl-2,3-dihydrobenzofuran-2-yl-1h-imidazole, 2-2-ethyl-2,3-dihydro-2-benzo b furanyl-1h-imidazole, 2-2-ethyl-2,3-dihydrobenzo b furan-2-yl-1h-imidazole
4302964
2-(2-ethyl-3H-1-benzofuran-2-yl)-1H-imidazole
CCC1(CC2=CC=CC=C2O1)C3=NC=CN3

Antagonist of pancreatic imidazoline receptor

Antagonist of the atypical imidazoline binding site (putative I3 receptor) of pancreatic β-cells. Selectively blocks efaroxan-induced insulin secretion in vitro and in vivo.
Quantity 10 mg
Formula Weight 214.27
Percent Purity >99%
Chemical Name or Material KU14R
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit