Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KU14R, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
189224-48-4
C13H14N2O
214.268
JCWVNNMJXQJVNC-UHFFFAOYSA-N
ku14r,2-2-ethyl-2,3-dihydro-2-benzofuranyl-1h-imidazole,2-2-ethyl-3h-1-benzofuran-2-yl-1h-imidazole,1h-imidazole,2-2-ethyl-2,3-dihydro-2-benzofuranyl,1h-imidazole, 2-2-ethyl-2,3-dihydro-2-benzofuranyl,ku-14r,2-2-ethyl-2,3-dihydrobenzofuran-2-yl-1h-imidazole,2-2-ethyl-2,3-dihydro-2-benzo b furanyl-1h-imidazole,2-2-ethyl-2,3-dihydrobenzo b furan-2-yl-1h-imidazole
4302964
2-(2-ethyl-3H-1-benzofuran-2-yl)-1H-imidazole
CCC1(CC2=CC=CC=C2O1)C3=NC=CN3
This item is not returnable. View return policy
189224-48-4
C13H14N2O
214.268
JCWVNNMJXQJVNC-UHFFFAOYSA-N
ku14r,2-2-ethyl-2,3-dihydro-2-benzofuranyl-1h-imidazole,2-2-ethyl-3h-1-benzofuran-2-yl-1h-imidazole,1h-imidazole,2-2-ethyl-2,3-dihydro-2-benzofuranyl,1h-imidazole, 2-2-ethyl-2,3-dihydro-2-benzofuranyl,ku-14r,2-2-ethyl-2,3-dihydrobenzofuran-2-yl-1h-imidazole,2-2-ethyl-2,3-dihydro-2-benzo b furanyl-1h-imidazole,2-2-ethyl-2,3-dihydrobenzo b furan-2-yl-1h-imidazole
4302964
2-(2-ethyl-3H-1-benzofuran-2-yl)-1H-imidazole
CCC1(CC2=CC=CC=C2O1)C3=NC=CN3

Antagonist of pancreatic imidazoline receptor

Antagonist of the atypical imidazoline binding site (putative I3 receptor) of pancreatic β-cells. Selectively blocks efaroxan-induced insulin secretion in vitro and in vivo.
Quantity 50mg
Formula Weight 214.27
Percent Purity >99%
Chemical Name or Material KU14R

Recommended Storage : Store at Room Temperature