Learn More
American Educational Products Kubic Bubbles Products for Science Education
Become entranced by these geometric bubbles.
Manufacturer:  American Educational Products RKB2
 View more versions of this product
Catalog No. S79051
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
Alert:
CHOKING HAZARD—Small parts. Not for children under 3 yrs.
Includes:
  • Instruction booklet
Description
Description
Build cubes, octahedrons, tetrahedrons or prisms with these plastic frames. Dip them into a soap-glycerin solution to create attention-getting shapes.
  • Illustrate light refraction, solutions to geometrical problems, surface areas, and properties of soap films
  • Can also be used with an overhead projector
Specifications
Specifications
To illustrate light refraction, solutions to geometrical problems, surface areas, and properties of soap films
Red, blue, green and gray tubes to make a cube, an octahedron, a prism and a tetrahedron, 2 perplex plates and 6 suction cups, instruction booklet
18
12
For overhead projector, secondary level
Kubic Bubbles Secondary Level Kit
12
7