Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kv1.2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP310294100UL

 View more versions of this product

Catalog No. NB125489


Add to cart

Description

Description

Kv1.2 Polyclonal antibody specifically detects Kv1.2 in Human samples. It is validated for Western Blot
Specifications

Specifications

Kv1.2
Unconjugated
PBS buffer, 2% sucrose
EC 3.6.1.27, EC 6.1.1, HBK5, HK4, HUKIV, Kv1.2, MGC50217, MK2, NGK1, potassium channel, potassium voltage-gated channel subfamily A member 2, potassium voltage-gated channel, shaker-related subfamily, member 2, RBK2, Voltage-gated K(+) channel HuKIV, Voltage-gated potassium channel HBK5, voltage-gated potassium channel protein Kv1.2, Voltage-gated potassium channel subunit Kv1.2
The immunogen is a synthetic peptide directed towards the N terminal region of human Kv1.2 (NP_001191198.1). Peptide sequence VATGDPADEAAALPGHPQDTYDPEADHECCERVVINISGLRFETQLKTLA
100 μg
Lipid and Metabolism, Neuroscience, Neurotransmission, Potassium Channels
Primary
3737
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Affinity purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit