Learn More
Kv1.4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 196971AP
Catalog No. 50-173-2494
Add to cart
Description
Description
The antibody is specific to KCN4.
Potassium voltage-gated channel subfamily A member 4, also known as Kv1.4 or PCN2, is a protein that in humans is encoded by the KCNA4 gene. This gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. It is mapped to 11p14.1. KCNA4 belongs to the A-type potassium current class, the members of which may be important in the regulation of the fast repolarizing phase of action potentials in heart and thus may influence the duration of cardiac action potential. KCNA4 also contributes to the cardiac transient outward potassium current (Ito1), the main contributing current to the repolarizing phase 1 of the cardiac action potential. This gene has been shown to interact with DLG4, KCNA2 and DLG1.
Specifications
Specifications
Kv1.4
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P15385, P22459, Q61423
HBK4, HK1, HPCN2, HUKII, KCNA4, KCNA4L, KCNA8, Kv1.4, PCN2
Rabbit
IgG
150 μL
-20°C
Primary
16492, 25469, 3739
Western Blot
Unconjugated
KCNA4
Liquid
Kcna4
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat