Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KV3.1 (KCNC1) Mouse anti-Human, Mouse, Rat, Clone: S16B-8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527684

Catalog No. PIMA527684


Add to cart

Description

Description

Voltage-gated K+ channels are important determits of neuronal membrane excitability. Moreover, differences in K+ channel expression patterns and densities contribute to the variations in action potential waveforms and repetitive firing patterns evident in different neuronal cell types. The Kv3.1 potassium channel is expressed at high levels in neurons that characteristically fire rapid trains of action potentials. Particularly high levels of this channel are found in neurons of the auditory brainstem. These neurons appear to participate in neural circuits that determine the intensity and timing of auditory stimuli and use this information to determine the location of sounds in space.
Specifications

Specifications

KV3.1 (KCNC1)
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
P15388, P25122, P48547
Krc2 1, NGK2, KCNC1, voltage gated potassium channel subunit Kv3.1, Kv4
Mouse
IgG1
100 μg
-20°C
Primary
16502, 25327, 3746
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
S16B-8
Unconjugated
KCNC1
Liquid
KCNC1
Fusion protein amino acids 437-585 (C-terminus) or rat Kv3.1b
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit