Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kv3.2 Antibody (S410-17), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259328B
Description
Kv3.2 Monoclonal specifically detects Kv3.2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Kv3.2 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
KCNC2 | |
Fusion protein amino acids 474-613 (Cytoplasmic C-terminus) of rat Kv3.2a | |
100 μL | |
Primary | |
Detects 80-100kDa. Varies due to post-translational modifications. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S410-17 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Kv3.2, MGC138196, potassium voltage-gated channel subfamily C member 2, potassium voltage-gated channel, Shaw-related subfamily, member 2, Voltage-gated potassium channel Kv3.2 | |
Mouse | |
Protein G purified | |
Signal Transduction | |
3747 | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction