Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kv3.2 Mouse anti-Human, Mouse, Rat, Clone: S410-17, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259328

Catalog No. NBP259328


Add to cart

Description

Description

Kv3.2 Monoclonal antibody specifically detects Kv3.2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Kv3.2
S410-17
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Purified
KCNC2
Fusion protein amino acids 474-613 (Cytoplasmic C-terminus) of rat Kv3.2a
Protein G purified
RUO
Primary
3747
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Unconjugated
PBS pH7.4, 50% glycerol with 0.1% Sodium Azide
Kv3.2, MGC138196, potassium voltage-gated channel subfamily C member 2, potassium voltage-gated channel, Shaw-related subfamily, member 2, Voltage-gated potassium channel Kv3.2
Mouse
IgG1
100 ug
Store at -20C.
Monoclonal
Detects 80-100kDa. Varies due to post-translational modifications.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only