Learn More
Kv3.2 Mouse anti-Human, Mouse, Rat, DyLight 680, Clone: S410-17, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259328FR
Catalog No. NBP259328FR
Add to cart
Description
Description
Kv3.2 Monoclonal antibody specifically detects Kv3.2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Kv3.2
S410-17
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
KCNC2
Fusion protein amino acids 474-613 (Cytoplasmic C-terminus) of rat Kv3.2a
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 80-100kDa. Varies due to post-translational modifications.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
Kv3.2, MGC138196, potassium voltage-gated channel subfamily C member 2, potassium voltage-gated channel, Shaw-related subfamily, member 2, Voltage-gated potassium channel Kv3.2
Mouse
IgG1
100 ul
Primary
3747
Human, Mouse, Rat