Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kv3.2 Mouse anti-Human,Mouse,Rat, Unconjugated, Clone: N410/17, MilliporeSigma™

Mouse Monoclonal Antibody

$184.00 - $463.00

Specifications

Antigen Kv3.2
Clone N410/17
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN24322MI
View Documents
MilliporeSigma
MABN243225UG
25 μg
Each for $184.00
Add to Cart
 
MABN24321MI
View Documents
MilliporeSigma
MABN2432100UG
100 μg
Each for $463.00
Add to Cart
 
Description

Description

Kv3.2 Monoclonal antibody specifically detects Kv3.2 in Human,Mouse,Rat samples. It is validated for Western Blotting, Immunohistochemistry (Paraffin).
Specifications

Specifications

Kv3.2
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human, Mouse, Rat
Potassium voltage-gated channel subfamily C member 2;Potassium channel voltage-gated Shaw-related subfamily C member 2;Shaw-like potassium channel;Voltage-gated potassium channel subunit Kv3.2.
A recombinant fragment corresponding to 140 amino acids from the C-terminal region of rat Voltage-gated potassium channel subunit Kv3.2.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
∼67 kDa observed; 67.55 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
N410/17
Monoclonal
Purified
Neuroscience
NP_631962
Kcnc2
IgG1 κ
Protein G Purified
Clone N410/17 is a mouse monoclonal antibody that specifically detects Voltage-gated potassium channel subunit Kv3.2. It targets an epitope with in the C-terminal region.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit