Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kv3.3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $320.71

Specifications

Antigen Kv3.3
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Applications Western Blot, Immunohistochemistry
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125614
View Documents
Novus Biologicals
NBP31035625UL
25 μg
Each for $106.19
Add to cart
 
NB125613
View Documents
Novus Biologicals
NBP310356100UL
100 μg
Each for $320.71
Add to cart
 
Description

Description

Kv3.3 Polyclonal antibody specifically detects Kv3.3 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

Kv3.3
Western Blot, Immunohistochemistry
Purified
RUO
Human
KSHIIID, KV3.3, potassium voltage-gated channel subfamily C member 3, potassium voltage-gated channel, Shaw-related subfamily, member 3, SCA13, Shaw-related voltage-gated potassium channel protein 3, spinocerebellar ataxia 13, voltage-gated potassium channel protein KV3.3, Voltage-gated potassium channel subunit Kv3.3
The immunogen is a synthetic peptide directed towards the middle region of human Kv3.3 (NP_004968). Peptide sequence YAERIGADPDDILGSNHTYFKNIPIGFWWAVVTMTTLGYGDMYPKTWSGM
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml, Immunohistochemistry
Unconjugated
Rabbit
Neuronal Cell Markers, Neuroscience, Neurotransmission
PBS buffer, 2% sucrose
3748
Polyclonal
Affinity purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit