Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kv3.4 Mouse anti-Human, Mouse, Rat, Biotin, Clone: S72-16, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147609B

Catalog No. NBP147609B


Add to cart

Description

Description

Kv3.4 Monoclonal specifically detects Kv3.4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Kv3.4
S72-16
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Q63734
HKSHIIIC, K+ channel subunit, KSHIIIC, Kv3.4, MGC126818, potassium voltage-gated channel subfamily C member 4, potassium voltage-gated channel, Shaw-related subfamily, member 4, Voltage-gated potassium channel subunit Kv3.4
Mouse
IgG1
0.1 ml
Primary
3749
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Biotin
PBS with 0.05% Sodium Azide
Purified
KCNC4
Synthetic peptide amino acids 175-192 of rat Kv3.4
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects approx 70kDa (100kDa in brain due to glycosylation).
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit