Learn More
Kv3.4 Mouse anti-Human, Mouse, Rat, DyLight 405, Clone: S72-16, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP147609V
Catalog No. NBP147609V
Add to cart
Description
Description
Kv3.4 Monoclonal specifically detects Kv3.4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
Kv3.4
S72-16
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Q63734
HKSHIIIC, K+ channel subunit, KSHIIIC, Kv3.4, MGC126818, potassium voltage-gated channel subfamily C member 4, potassium voltage-gated channel, Shaw-related subfamily, member 4, Voltage-gated potassium channel subunit Kv3.4
Mouse
IgG1
0.1 ml
Primary
3749
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Purified
KCNC4
Synthetic peptide amino acids 175-192 of rat Kv3.4
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects approx 70kDa (100kDa in brain due to glycosylation).