Learn More
Kv3.4 Mouse anti-Human, Mouse, Rat, HRP, Clone: S72-16, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP147609H
Catalog No. NBP147609H
Add to cart
Description
Description
Kv3.4 Monoclonal antibody specifically detects Kv3.4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Kv3.4
S72-16
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Q63734
HKSHIIIC, K+ channel subunit, KSHIIIC, Kv3.4, MGC126818, potassium voltage-gated channel subfamily C member 4, potassium voltage-gated channel, Shaw-related subfamily, member 4, Voltage-gated potassium channel subunit Kv3.4
Mouse
IgG1
0.1 ml
Primary
3749
Human, Mouse, Rat
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
HRP
PBS with No Preservative
Purified
KCNC4
Synthetic peptide amino acids 175-192 of rat Kv3.4
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects approx 70kDa (100kDa in brain due to glycosylation).