Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KV4.3 (KCND3) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541026
Description
Peptide sequence: KARLARIRVA KTGSSNAYLH SKRNGLLNEA LELTGTPEEE HMGKTTSLIE Sequence homology: Cow: 92%; Dog: 92%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). This gene encodes a member of the potassium channel, voltage-gated, shal-related subfamily, members of which form voltage-activated A-type potassium ion channels and are prominent in the repolarization phase of the action potential. This member includes two isoforms with different sizes, which are encoded by alternatively spliced transcript variants of this gene.
Specifications
KV4.3 (KCND3) | |
Polyclonal | |
Unconjugated | |
KCND3 | |
AW045978; BRGDA9; KCND3; KCND3L; KCND3S; Kncd3; KSHIVB; KV4.3; potassium channel alpha subunit Kv4.3 short form; potassium channel Kv4.3; potassium channel Kv4.3L; potassium channel Kv4.3M; potassium channel, voltage gated Shal related subfamily D, member 3; potassium channel, voltage-gated Shal-related subfamily D, member 3; potassium ionic channel Kv4.3; potassium voltage gated channel, Shal-related family, member 3; potassium voltage-gated channel long; potassium voltage-gated channel subfamily D member 3; potassium voltage-gated channel, Shal-related family, member 3; potassium voltage-gated channel, Shal-related subfamily, member 3; SCA19; SCA22; sha1-related potassium channel Kv4.3; voltage-gated K+ channel; voltage-gated potassium channel subunit Kv4.3 | |
Rabbit | |
Protein A | |
RUO | |
3752, 65195 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q62897, Q9UK17 | |
KCND3 | |
Synthetic peptide directed towards the middle region of human KCND3. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction