Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kv9.1 Rabbit, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP18006420UL

 View more versions of this product

Catalog No. NBP18006420


Add to cart

Description

Description

Kv9.1 Polyclonal antibody specifically detects Antigen in Human, Primate samples. It is validated for Western Blotting.
Specifications

Specifications

Kv9.1
Unconjugated
PBS & 2% Sucrose. with No Preservative
Affinity Purified
KCNS1
Synthetic peptide directed towards the N terminal of human KCNS1. Peptide Sequence: LCDDYDEAAREFYFDRHPGFFLSLLHFYRTGHLHVLDELCVFAFGQEADY
58 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human, Primate
Western Blot
Western Blot 1:1000
NP_002242
potassium voltage-gated channel subfamily S member 1, potassium voltage-gated channel, delayed-rectifier, subfamily S, member 1
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
3787
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit