Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kv9.2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Kv9.2 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Kv9.2 Polyclonal specifically detects Kv9.2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Kv9.2 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
3788 | |
IgG | |
Affinity purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Delayed-rectifier K(+) channel alpha subunit 2, KIAA1144, Kv9.2, potassium voltage-gated channel subfamily S member 2, potassium voltage-gated channel, delayed-rectifier, subfamily S, member 2, Voltage-gated potassium channel subunit Kv9.2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-187 of human Kv9.2 (NP_065748.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title