Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kv9.3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303307100UL
Description
Kv9.3 Polyclonal specifically detects Kv9.3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Kv9.3 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
3790 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry | |
Unconjugated | |
delayed-rectifier K(+) channel alpha subunit 3, KV9.3, MGC9481, potassium voltage-gated channel subfamily S member 3, potassium voltage-gated channel, delayed-rectifier, subfamily S, member 3, Shab-related delayed-rectifier K+ channel alpha subunit 3, voltage-gated potassium channel protein Kv9.3, voltage-gated potassium channel subunit Kv9.3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human Kv9.3 (NP_002243.3). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction