Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KvBeta1 Antibody (OTI7F12), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271349PECY55
Description
KvBeta1 Monoclonal specifically detects KvBeta1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
KvBeta1 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
KCNAB1 | |
Full length recombinant protein of human KvBeta1 (NP_751891) produced in HEK293T cells. | |
0.1 mL | |
7881.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI7F12 | |
Flow Cytometry | |
AKR6A3, hKvb3, hKvBeta3, K(+) channel subunit beta-1, KCNA1Bpotassium channel beta 3 chain, Kvb1.3, KV-BETA-1, potassium channel beta3 subunit, potassium channel shaker chain beta 1a, potassium voltage-gated channel beta subunit, potassium voltage-gated channel, shaker-related subfamily, beta member 1, voltage-gated potassium channel beta-1 subunit, voltage-gated potassium channel subunit beta-1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction