Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KY 02111, Tocris Bioscience™

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
1118807-13-8
C18H17ClN2O3S
376.855
LXFKEVQQSKQXPR-UHFFFAOYSA-N
n-6-chlorobenzo d thiazol-2-yl-3-3,4-dimethoxyphenyl propanamide,n-6-chloro-1,3-benzothiazol-2-yl-3-3,4-dimethoxyphenyl propanamide,n-6-chloro-2-benzothiazolyl-3,4-dimethoxybenzenepropanamide,benzenepropanamide, n-6-chloro-2-benzothiazolyl-3,4-dimethoxy,n-6-chloro-benzothiazol-2-yl-3-3,4-dimethoxy-phenyl-propionamide
8582409
N-(6-chloro-1,3-benzothiazol-2-yl)-3-(3,4-dimethoxyphenyl)propanamide
COC1=C(C=C(C=C1)CCC(=O)NC2=NC3=C(S2)C=C(C=C3)Cl)OC
This item is not returnable. View return policy
1118807-13-8
C18H17ClN2O3S
376.855
LXFKEVQQSKQXPR-UHFFFAOYSA-N
n-6-chlorobenzo d thiazol-2-yl-3-3,4-dimethoxyphenyl propanamide,n-6-chloro-1,3-benzothiazol-2-yl-3-3,4-dimethoxyphenyl propanamide,n-6-chloro-2-benzothiazolyl-3,4-dimethoxybenzenepropanamide,benzenepropanamide, n-6-chloro-2-benzothiazolyl-3,4-dimethoxy,n-6-chloro-benzothiazol-2-yl-3-3,4-dimethoxy-phenyl-propionamide
8582409
N-(6-chloro-1,3-benzothiazol-2-yl)-3-(3,4-dimethoxyphenyl)propanamide
COC1=C(C=C(C=C1)CCC(=O)NC2=NC3=C(S2)C=C(C=C3)Cl)OC

Inhibits canonical Wnt signaling. Promotes differentiation of human ESCs and iPSCs into cardiomyocytes

Promotes differentiation of human ESCs and iPSCs into cardiomyocytes when used after day three in culture; differentiation efficiency increases when used after initial culture with CHIR 99021 (Cat. No. 4423). Induces downregulation of Wnt signaling target genes; inhibits canonical Wnt signaling in a manner distinct from other known Wnt inhibitors.
Quantity 10mg
Formula Weight 376.86
Percent Purity >99%
Chemical Name or Material KY 02111

Recommended Storage : Store at +4°C