Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kynureninase Antibody (OTI1H1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245579
Description
Ensure accurate, reproducible results in Immunohistochemistry
Kynureninase Monoclonal specifically detects Kynureninase in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Kynureninase | |
Monoclonal | |
1 mg/mL | |
Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
EC 3.7.1.3, kynureninase, kynureninase (L-kynurenine hydrolase), L-kynurenine hydrolase | |
Mouse | |
34.5 kDa | |
0.1 mL | |
Primary | |
Human, Rat | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI1H1 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
KYNU | |
Human recombinant protein fragment corresponding to amino acids 1-216 of human KYNU (NP_001028170) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
8942 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction