Learn More
Novus Biologicals™ Kynurenine 3-Monooxygenase/KMO Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP186335PEP
Catalog No. NBP86335PEP
Add to cart
Description
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human KMO. The Kynurenine 3-Monooxygenase/KMO Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
Human
>80% by SDS-PAGE and Coomassie blue staining
100μL
E. coli
Store at -20°C. Avoid freeze-thaw cycles.
kynurenine 3-monooxygenase (kynurenine 3-hydroxylase)
31 kDa
8564
Antibody Competition
PBS and 1M Urea, pH 7.4.
No Preservative
KMO