Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-165,041, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent PPARδ agonist (Ki = 6 nM); displays > 100-fold selectivity for both mouse and human PPARδ receptors over other subtypes. Raises plasma cholesterol levels in vivo, in insulin-resistant db/db mice. Neuroprotective in models of cerebral infarction and Parkinson's disease.
Specifications
Quantity | 10 mg |
Formula Weight | 402.44 |
Percent Purity | >98% |
Chemical Name or Material | L-165, 041 |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction