Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(-)-2-Amino-3-phenyl-1-propanol, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
3182-95-4
C9H13NO
151.21
MFCD00004732
STVVMTBJNDTZBF-VIFPVBQESA-N
l-phenylalaninol,s-2-amino-3-phenyl-1-propanol,s---2-amino-3-phenyl-1-propanol,s-2-amino-3-phenylpropan-1-ol,h-phenylalaninol,h-phe-ol,2s-2-amino-3-phenylpropan-1-ol,phenylalaninol,--l-phenylalaninol,l-phenyl alaninol
447213
(2S)-2-amino-3-phenylpropan-1-ol
C1=CC=C(C=C1)CC(CO)N
3182-95-4
C9H13NO
151.21
MFCD00004732
STVVMTBJNDTZBF-VIFPVBQESA-N
l-phenylalaninol,s-2-amino-3-phenyl-1-propanol,s---2-amino-3-phenyl-1-propanol,s-2-amino-3-phenylpropan-1-ol,h-phenylalaninol,h-phe-ol,2s-2-amino-3-phenylpropan-1-ol,phenylalaninol,--l-phenylalaninol,l-phenyl alaninol
447213
(2S)-2-amino-3-phenylpropan-1-ol
C1=CC=C(C=C1)CC(CO)N
Specific Rotation -23
Quantity 5g
Melting Point 90°C to 94°C
Color White to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula C6H5CH2CH(NH2)CH2OH
Solubility Information Solubility in water: soluble. Other solubilities: very soluble in ethanol,,soluble in chloroform
Formula Weight 151.21
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material L(-)-2-Amino-3-phenyl-1-propanol
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 221-674-4