Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ L-(+)-2-Phenylglycine, 98+%
Click to view available options
Quantity:
25g
100g
500g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Beilstein | 2208675 |
Merck Index | 14,7291 |
Formula Weight | 151.17 |
Percent Purity | ≥98% |
Melting Point | 288°C (decomposition) |
Quantity | 100g |
Optical Rotation | +156° (c=1 in 1N HCl) |
Chemical Name or Material | L-(+)-2-Phenylglycine |
Safety and Handling
EINECSNumber : 220-909-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only