Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride, 95%, Acros Organics

Click to view available options
Quantity:
1g
5g
1421-65-4
C10H13NO4·ClH
247.68
WFGNJLMSYIJWII-FJXQXJEOSA-N
s-methyl 2-amino-3-3,4-dihydroxyphenyl propanoate hydrochloride,l-3,4-dihydroxyphenylalanine methyl ester hydrochloride,methyl l-dopa hydrochloride,melevodopa hydrochloride,unii-jfu1a8866v,l-dopa methyl ester hydrochloride,levodopa methyl ester hydrochloride,l-tyrosine, 3-hydroxy-, methyl ester, hydrochloride,dsstox_cid_25763,dsstox_rid_81105
10131132
methyl (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoate;hydrochloride
COC(=O)C(CC1=CC(=C(C=C1)O)O)N.Cl
1421-65-4
C10H13NO4·ClH
247.68
WFGNJLMSYIJWII-FJXQXJEOSA-N
s-methyl 2-amino-3-3,4-dihydroxyphenyl propanoate hydrochloride,l-3,4-dihydroxyphenylalanine methyl ester hydrochloride,methyl l-dopa hydrochloride,melevodopa hydrochloride,unii-jfu1a8866v,l-dopa methyl ester hydrochloride,levodopa methyl ester hydrochloride,l-tyrosine, 3-hydroxy-, methyl ester, hydrochloride,dsstox_cid_25763,dsstox_rid_81105
10131132
methyl (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoate;hydrochloride
COC(=O)C(CC1=CC(=C(C=C1)O)O)N.Cl
Packaging Glass Bottle
Melting Point 94.2°C to 94.3°C
Quantity 1g
Specific Rotation +9.0° to +11.0° (20°C, 589nm) (C=2 in methanol)
Elemental Analysis 45.58 to 51.40 % (C) Dry basis 5.32 to 5.99 % (N) Dry basis
Water 1% Max.
Infrared Spectrum Authentic
Color White
Assay Percent Range 95%
Formula Weight 247.68
Physical Form Powder or Crystalline Powder
Percent Purity 95%
Chemical Name or Material L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning