Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-651,582, Tocris Bioscience™

Catalog Number p-4784081
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
99519-84-3
C17H12Cl3N5O2
424.67
MFCD00866325
WNRZHQBJSXRYJK-UHFFFAOYSA-N
carboxyamidotriazole, carboxyamido-triazole, 5-amino-1-3,5-dichloro-4-4-chlorobenzoyl benzyl-1h-1,2,3-triazole-4-carboxamide, cai, 4-cai, unii-6st3zf52wb, 6st3zf52wb, 5-amino-1-3,5-dichloro-4-4-chlorobenzoyl phenyl methyl triazole-4-carboxamide, 1h-1,2,3-triazole-4-carboxamide, 5-amino-1-3,5-dichloro-4-4-chlorobenzoyl phenyl methyl, 5-amino-1-3,5-dichloro-4-4-chlorobenzoyl phenyl methyl-1h-1,2,3-triazole-4-carboxamide
108144
5-amino-1-{[3,5-dichloro-4-(4-chlorobenzoyl)phenyl]methyl}-1H-1,2,3-triazole-4-carboxamide
NC(=O)C1=C(N)N(CC2=CC(Cl)=C(C(=O)C3=CC=C(Cl)C=C3)C(Cl)=C2)N=N1
This item is not returnable. View return policy
99519-84-3
C17H12Cl3N5O2
424.67
MFCD00866325
WNRZHQBJSXRYJK-UHFFFAOYSA-N
carboxyamidotriazole, carboxyamido-triazole, 5-amino-1-3,5-dichloro-4-4-chlorobenzoyl benzyl-1h-1,2,3-triazole-4-carboxamide, cai, 4-cai, unii-6st3zf52wb, 6st3zf52wb, 5-amino-1-3,5-dichloro-4-4-chlorobenzoyl phenyl methyl triazole-4-carboxamide, 1h-1,2,3-triazole-4-carboxamide, 5-amino-1-3,5-dichloro-4-4-chlorobenzoyl phenyl methyl, 5-amino-1-3,5-dichloro-4-4-chlorobenzoyl phenyl methyl-1h-1,2,3-triazole-4-carboxamide
108144
5-amino-1-{[3,5-dichloro-4-(4-chlorobenzoyl)phenyl]methyl}-1H-1,2,3-triazole-4-carboxamide
NC(=O)C1=C(N)N(CC2=CC(Cl)=C(C(=O)C3=CC=C(Cl)C=C3)C(Cl)=C2)N=N1

Ca2+ channel blocker

Orally active Ca2+ channel blocker; inhibits K+- and carbachol-stimulated Ca2+ influx (IC50 values are 500 and 935 nM respectively). Exhibits antiproliferative, antiangiogenic and antimetastatic activity in vivo and displays selectivity towards numerous mismatch repair-deficient tumor cell lines in vitro.
Quantity 10 mg
Formula Weight 424.67
Percent Purity >98%
Chemical Name or Material L-651, 582
Recommended Storage Store at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit