Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-670,596, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
121083-05-4
C22H21F2NO4S
433.47
GMJWAIMJHBTYPD-UHFFFAOYSA-N
--6-8-difluoro-2,3,4,9-tetrahydro-9-4-methylsulfonyl phenyl methyl-1h-carbazole-1-acetic acid,d0an1y,6,8-difluoro-9-4-methanesulfonylphenyl methyl-1,2,3,4-tetrahydrocarbazol-1-yl acetic acid,1h-carbazole-1-acetic acid, 6,8-difluoro-2,3,4,9-tetrahydro-9-4-methylsulfonyl phenyl methyl-,-,1h-carbazole-1-aceticacid,6,8-difluoro-2,3,4,9-tetrahydro-9-4-methylsulfonyl phenyl methyl-,-,2-6,8-difluoro-9-4-methylsulfonyl benzyl-2,3,4,9-tetrahydro-1h-carbazol-1-yl acetic acid,2-6,8-difluoro-9-4-methylsulfonylphenyl methyl-1,2,3,4-tetrahydrocarbazol-1-yl acetic acid
129360
2-[6,8-difluoro-9-[(4-methylsulfonylphenyl)methyl]-1,2,3,4-tetrahydrocarbazol-1-yl]acetic acid
CS(=O)(=O)C1=CC=C(C=C1)CN2C3=C(CCCC3CC(=O)O)C4=CC(=CC(=C42)F)F
This item is not returnable. View return policy
121083-05-4
C22H21F2NO4S
433.47
GMJWAIMJHBTYPD-UHFFFAOYSA-N
--6-8-difluoro-2,3,4,9-tetrahydro-9-4-methylsulfonyl phenyl methyl-1h-carbazole-1-acetic acid,d0an1y,6,8-difluoro-9-4-methanesulfonylphenyl methyl-1,2,3,4-tetrahydrocarbazol-1-yl acetic acid,1h-carbazole-1-acetic acid, 6,8-difluoro-2,3,4,9-tetrahydro-9-4-methylsulfonyl phenyl methyl-,-,1h-carbazole-1-aceticacid,6,8-difluoro-2,3,4,9-tetrahydro-9-4-methylsulfonyl phenyl methyl-,-,2-6,8-difluoro-9-4-methylsulfonyl benzyl-2,3,4,9-tetrahydro-1h-carbazol-1-yl acetic acid,2-6,8-difluoro-9-4-methylsulfonylphenyl methyl-1,2,3,4-tetrahydrocarbazol-1-yl acetic acid
129360
2-[6,8-difluoro-9-[(4-methylsulfonylphenyl)methyl]-1,2,3,4-tetrahydrocarbazol-1-yl]acetic acid
CS(=O)(=O)C1=CC=C(C=C1)CN2C3=C(CCCC3CC(=O)O)C4=CC(=CC(=C42)F)F

Potent, selective thromboxane A2/prostaglandin endoperoxide antagonist

Potent and selective thromboxane A2/prostaglandin endoperoxide receptor antagonist (IC50 = 5.5 nM). Inhibits U-44069-induced contractions of guinea pig trachea (pA2 = 9.0) and human platelet aggregation in vitro (IC50 = 6.5 nM). Also prevents thromboxane-mediated endothelial cell death. Orally active in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 433.47
Percent Purity >98%
Chemical Name or Material L-670, 596

Recommended Storage : Store at Room Temperature