Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-694,247, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

137403-12-4
C20H21N5O3S
411.48
HKXMQLISPYELRD-UHFFFAOYSA-N
2-[5-[3-(4-Methylsulfonylamino)benzyl-1, 2, 4-oxadiazol-5-yl]-1H-indol-3-yl]ethanamine
132059

5-HT1D agonist

Selective and extremely potent 5-HT1D receptor agonist.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
SMILES CS(=O)(=O)NC1=CC=C(C=C1)CC2=NOC(=N2)C3=CC4=C(C=C3)NC=C4CCN
IUPAC Name N-[4-[[5-[3-(2-aminoethyl)-1H-indol-5-yl]-1,2,4-oxadiazol-3-yl]methyl]phenyl]methanesulfonamide
Formula Weight 411.48
Chemical Name or Material L-694, 247
Grade >98%

Recommended Storage : Desiccate at +4°C