Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-694,247, Tocris Bioscience™

Catalog Number p-4783570
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
137403-12-4
C20H21N5O3S
411.48
HKXMQLISPYELRD-UHFFFAOYSA-N
n-4-5-3-2-aminoethyl-1h-indol-5-yl-1,2,4-oxadiazol-3-yl methyl phenyl methanesulfonamide, 2-5-3-4-methylsulfonylamino benzyl-1,2,4-oxadiazol-5-yl-1h-indol-3-yl ethanamine, methanesulfonamide,n-4-5-3-2-aminoethyl-1h-indol-5-yl-1,2,4-oxadiazol-3-yl methyl phenyl, tocris-0781, biomol-nt_000112, d0g1rt, gtpl15, 2-5-3-4-methylsulfonylamino benzyl-1,2,4-oxadiazol-5-yl-1h-indole-3-yl ethylamine
132059
N-[4-[[5-[3-(2-aminoethyl)-1H-indol-5-yl]-1,2,4-oxadiazol-3-yl]methyl]phenyl]methanesulfonamide
CS(=O)(=O)NC1=CC=C(C=C1)CC2=NOC(=N2)C3=CC4=C(C=C3)NC=C4CCN
This item is not returnable. View return policy
137403-12-4
C20H21N5O3S
411.48
HKXMQLISPYELRD-UHFFFAOYSA-N
n-4-5-3-2-aminoethyl-1h-indol-5-yl-1,2,4-oxadiazol-3-yl methyl phenyl methanesulfonamide, 2-5-3-4-methylsulfonylamino benzyl-1,2,4-oxadiazol-5-yl-1h-indol-3-yl ethanamine, methanesulfonamide,n-4-5-3-2-aminoethyl-1h-indol-5-yl-1,2,4-oxadiazol-3-yl methyl phenyl, tocris-0781, biomol-nt_000112, d0g1rt, gtpl15, 2-5-3-4-methylsulfonylamino benzyl-1,2,4-oxadiazol-5-yl-1h-indole-3-yl ethylamine
132059
N-[4-[[5-[3-(2-aminoethyl)-1H-indol-5-yl]-1,2,4-oxadiazol-3-yl]methyl]phenyl]methanesulfonamide
CS(=O)(=O)NC1=CC=C(C=C1)CC2=NOC(=N2)C3=CC4=C(C=C3)NC=C4CCN

5-HT1D agonist

Selective and extremely potent 5-HT1D receptor agonist.
Quantity 10 mg
Formula Weight 411.48
Percent Purity >98%
Chemical Name or Material L-694, 247
Recommended Storage Desiccate at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit