Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-694,247, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
137403-12-4
C20H21N5O3S
411.48
HKXMQLISPYELRD-UHFFFAOYSA-N
n-4-5-3-2-aminoethyl-1h-indol-5-yl-1,2,4-oxadiazol-3-yl methyl phenyl methanesulfonamide,2-5-3-4-methylsulfonylamino benzyl-1,2,4-oxadiazol-5-yl-1h-indol-3-yl ethanamine,methanesulfonamide,n-4-5-3-2-aminoethyl-1h-indol-5-yl-1,2,4-oxadiazol-3-yl methyl phenyl,n-4-5-3-2-aminoethyl-1h-indol-5-yl-1,2,4-oxadiazol-3-yl methyl phenyl methanesulfonamide,n-4-5-3-2-aminoethyl-1h-indol-5-yl-1,2,4-oxadiazol-3-yl methyl phenyl methanesulfonamide,tocris-0781,biomol-nt_000112,d0g1rt,gtpl15,2-5-3-4-methylsulfonylamino benzyl-1,2,4-oxadiazol-5-yl-1h-indole-3-yl ethylamine
132059
N-[4-[[5-[3-(2-aminoethyl)-1H-indol-5-yl]-1,2,4-oxadiazol-3-yl]methyl]phenyl]methanesulfonamide
CS(=O)(=O)NC1=CC=C(C=C1)CC2=NOC(=N2)C3=CC4=C(C=C3)NC=C4CCN
137403-12-4
C20H21N5O3S
411.48
HKXMQLISPYELRD-UHFFFAOYSA-N
n-4-5-3-2-aminoethyl-1h-indol-5-yl-1,2,4-oxadiazol-3-yl methyl phenyl methanesulfonamide,2-5-3-4-methylsulfonylamino benzyl-1,2,4-oxadiazol-5-yl-1h-indol-3-yl ethanamine,methanesulfonamide,n-4-5-3-2-aminoethyl-1h-indol-5-yl-1,2,4-oxadiazol-3-yl methyl phenyl,n-4-5-3-2-aminoethyl-1h-indol-5-yl-1,2,4-oxadiazol-3-yl methyl phenyl methanesulfonamide,n-4-5-3-2-aminoethyl-1h-indol-5-yl-1,2,4-oxadiazol-3-yl methyl phenyl methanesulfonamide,tocris-0781,biomol-nt_000112,d0g1rt,gtpl15,2-5-3-4-methylsulfonylamino benzyl-1,2,4-oxadiazol-5-yl-1h-indole-3-yl ethylamine
132059
N-[4-[[5-[3-(2-aminoethyl)-1H-indol-5-yl]-1,2,4-oxadiazol-3-yl]methyl]phenyl]methanesulfonamide
CS(=O)(=O)NC1=CC=C(C=C1)CC2=NOC(=N2)C3=CC4=C(C=C3)NC=C4CCN

5-HT1D agonist

Selective and extremely potent 5-HT1D receptor agonist.
Quantity 50mg
Formula Weight 411.48
Percent Purity >98%
Chemical Name or Material L-694, 247

Recommended Storage : Desiccate at +4°C